MOPS-SDS running buffer (20X)
- Fyzická forma:Liquid
Frequently Bought Together
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Agar for bacteriology
: VWR Chemicals
Aceton ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Bags, zip seal, Minigrip®
: Flexico