Anti-CCAR2 Goat Polyclonal Antibody
Catalog # EVBIEB12588
Supplier: Everest Biotech
Specifications
- Druh protilátky:Primární
- Název antigenu:Cell cycle and apoptosis regulator 2
- Symbol antigenu:CCAR2
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Goat
- Izotyp:IgG
- Reaktivita:Pig,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Formát:affinity purified
- Formulář:liquid
- Identifikační číslo genu:57805
- Synonyma antigenu:p30DBC|NET35|KIAA1967|DBC-1|p30 DBC|DBC1
- Skladovací pufr:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sekvence:C-ESHNRFSATEVTNK
- Skladovací teplota:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Koncentrace:100µg/200µl
- Transportní teplota:8 °C
- Čištění:Affinity purified
- Rozměry protilátky:100µg
- Typ balení:plastic vial
- Bal.:0,1 mg
Specifications
About this item
Western Blot: Approx 140kDa band observed in nuclear lysates of cell lines HEK293 and MCF7 (calculated MW of 103kDa according to NP_066997.3). The observed molecular weight is explained by its acetylation (Zheng et al, Mol Cell Biol. 2013 Dec;33(24):496
Type: Primary
Antigen: CCAR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog, Pig, Bovine