Anti-CSF3R Goat Polyclonal Antibody
Katalogové číslo EVBIEB11702
Supplier: Everest Biotech
Specifications
- Druh protilátky:Primární
- Název antigenu:colony stimulating factor 3 receptor (granulocyte)
- Symbol antigenu:CSF3R
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Goat
- Izotyp:IgG
- Reaktivita:Human,Bovine
- Western blot:Yes
- Formát:affinity purified
- Formulář:liquid
- Identifikační číslo genu:1441
- Synonyma antigenu:GCSFR|CD114
- Skladovací pufr:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
- Sekvence:C-PPELGKSPLTHYT
- Skladovací teplota:Store at -20° C prior to opening. Aliquot contents and freeze at -20° C or below for extended storage. Avoid cycles of freezing and thawing
- Koncentrace:100µg/200µl
- Transportní teplota:8 °C
- Čištění:Affinity purified
- Rozměry protilátky:100µg
- Typ balení:plastic vial
- Bal.:0,1 mg
Specifications
About this item
Western Blot: Approx 100kDa band observed in Human Peripheral Blood Mononucleocytes lysates (calculated MW of 92.1kDa according to NP_000751.1). The observed molecular weight corresponds to the glycosylated form. Recommended concentration: 1-3µg/ml.
Type: Primary
Antigen: CSF3R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Bovine