ContactUsLinkComponent
Horizontal electrophoresis system, multiSUB midi
Horizontal electrophoresis system, multiSUB midi
Supplier:  CLEAVER SCIENTIFIC
Certificates
undefined
Horizontal electrophoresis system, multiSUB midi
Supplier:  CLEAVER SCIENTIFIC
Specifications
  • Catalog No:
  • CLEVMSMIDI7
  • CLEVMSMIDIDUO
  • Model:
  • multiSUB midi 7
  • multiSUB midi DUO
  • Velikost gelu Š x D:
  • 100×70 mm
  • 100×70 ; 100×100 mm
  • Objem vzorku:
  • 50 samples
  • Objem pufru:
  • 300 ml
  • 300 ml
  • ŠxDxV přístroje:
  • 125×220×90 mm
  • 125×220×90 mm