Test kits, oxygen scavengers test, Spectroquant®, Supelco®
1.19251.0001
: Merck
:
- Chemická metoda:Iron reduction
- Bal.:200 Tests
- Popis:Oxygen scavengers test
- M-Clarity quality level:MQ100
- Pro jednotku:Prove, Nova 60, Multy
- Rozsah:0,020 - 0,500 mg/l DEHA
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Highly stable, ready to use reagent mixtures in a user friendly design
- Free selection of appropriate cell format according to the measuring range
- Long shelf life up to 3 years - all at room temperature
- Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.