Test kits, acid capacity (total alkalinity), Spectroquant®, Supelco®
Katalogové číslo 1.01758.0001
Dodavatel: Merck
Číslo CAS:
Specifikace
- Chemická metoda:Indicator
- Bal.:120 Tests
- Popis:Acid capacity cell test to pH 4,3 (total alkalinity)
- Pro jednotku:Prove, Nova 30/60, Multy, Move
- Rozsah:20 - 400 mg/l CaCO₃
Specifikace
O tomto produktu
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used, not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
- Very user friendly, validated methods
- All necessary reagents for the analysis are contained in the cells for most test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.