Anti-CD55 Mouse Monoclonal Antibody [clone: 561CT4.2.3]
Catalog # ABGEAM2092B
Supplier: Abgent
Specifications
- Druh protilátky:Primární
- Název antigenu:CD55 molecule, decay accelerating factor for complement (Cromer blood group)
- Symbol antigenu:CD55
- Klonovalita:Monoclonal
- Klon:561CT4.2.3
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Mouse
- Izotyp:IgG1
- Reaktivita:Human
- Western blot:Yes
- Identifikační číslo genu:1604
- Synonyma antigenu:CR|DAF|CROM|TC
- Počet aminokyselin:51 to 79
- Skladovací pufr:pH 7,4
- Molekulární hmotnost:41.4 kDa
- Sekvence:EESFVKIPGEKDSVC
- Skladovací teplota:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Koncentrace:0.4
- Transportní teplota:4 °C
- Imunogen:This CD55 antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 51-79 amino acids from human CD55.
- Čištění:Protein G Purification
- Bal.:400 µl
Specifications
About this item
Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: 561CT4.2.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human