Dropping bottles, amber
Dodavatel: Lokální dodavatel
Přejít na:
O tomto produktu
Glass, amber.
- Good chemical and physical properties
- For use in applications that do not involve high thermal stresses
- Screw cap with bulb and pipette must be ordered seperately
Amber to protect from UV rays and light-sensitive substances.
2 Dostupných variant níže
Product Details & Documents
Glass, amber.
- Good chemical and physical properties
- For use in applications that do not involve high thermal stresses
- Screw cap with bulb and pipette must be ordered seperately
Amber to protect from UV rays and light-sensitive substances.
Product Family Options
Informace o produktu
- SekvenceKapacitaDostupnostPrice
This product has been blocked by your organization. Please contact your purchasing department for more information
Accessories
Informace o produktu
- Katalogové čísloDostupnostPrice
- Screw cap with gasket for dropping bottles
Informace o produktu
- Katalogové čísloDostupnostPrice
- Screw cap with inner dropping stopper
Informace o produktu
- Katalogové čísloDostupnostPrice
- Screw cap with pipette and bulb for10 ml dropping bottlesScrew cap with pipette and bulb for10 ml dropping bottlesVITU50122Přihlásit se pro dostupnost1 * 1 KS - 19,00 Kč
Informace o produktu
- Katalogové čísloDostupnostPrice
- Screw cap with pipette and bulb for 20 ml dropping bottlesScrew cap with pipette and bulb for 20 ml dropping bottlesVITU50124Přihlásit se pro dostupnost1 * 1 KS - 19,40 Kč