Anti-EPS8L2 Rabbit Polyclonal Antibody
Catalog # ABGEAP19537A
Supplier: Abgent
Specifications
- Druh protilátky:Primární
- Název antigenu:EPS8-like 2
- Symbol antigenu:EPS8L2
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Reaktivita:Human
- Western blot:Yes
- Epitop:N-terminal
- Identifikační číslo genu:64787
- Synonyma antigenu:EPS8R2
- Počet aminokyselin:162 to 190
- Skladovací pufr:pH 7,4
- Molekulární hmotnost:80.621 kDa
- Sekvence:GKKMRPQTLKGHQEC
- Skladovací teplota:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Koncentrace:0.47
- Transportní teplota:4 °C
- Imunogen:This EPS8L2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 162-190 amino acids from the N-terminal region of human EPS8L2.
- Čištění:Protein A & Peptide affinity purification
- Bal.:400 µl
Specifications
About this item
Type: Primary
Antigen: EPS8L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human