Anti-WRNIP1 Rabbit Polyclonal Antibody
ABGEAP19098A
: Abgent
- Druh protilátky:Primární
- Název antigenu:Werner helicase interacting protein 1
- Symbol antigenu:WRNIP1
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Rabbit
- Imunochemie:Yes
- Reaktivita:Human
- Western blot:Yes
- Epitop:N-terminal
- Identifikační číslo genu:56897
- Synonyma antigenu:bA420G6.2|WHIP
- Počet aminokyselin:1 to 30
- Skladovací pufr:pH 7,4
- Molekulární hmotnost:72.133 kDa
- Sekvence:EVSGPEDDPFLSC
- Skladovací teplota:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Koncentrace:0.6
- Transportní teplota:4 °C
- Imunogen:This WRNIP1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human WRNIP1.
- Čištění:Protein A & Peptide affinity purification
- Bal.:400 µl
Type: Primary
Antigen: WRNIP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human