Visual colour kits, nickel, MQuant®, Supelco®
Catalog # 1.18461.0002
Supplier: Merck
CAS Number:
Specifications
- Chemická metoda:Dimethylglyoxime
- Bal.:500 Tests
- Popis:Nickel test, RP for 1.14783.0001; 1.14420.0001
- Sekvence:04
- M-Clarity quality level:MQ100
- Typ:MCD
Specifications
About this item
Quick tests for visual colour comparison.
- Results in approximately 5 minutes
- Easy to use
- Application-oriented measuring ranges
- Portable
- A range of economical refill packs (RP) are available
MQuant® with colour disc comparator
Using the comparator disc, these test kits are especially suitable for the analysis of turbid and coloured waters. The comparator disc is a practical and robust colour comparison device that provides reliable measurements, even in humid environments. Ideal for samples of medium to high concentration in applications ranging from testing bottled water to wastewater and other industrial water testing.
RP= Refill pack
Delivery information: Each test kit contains all the necessary reagents, reaction cuvettes, colour comparison chart or comparator wheel and extensive instructions for use.