Hydrazine colour disc test kit, HY-2
HACH184900
: Hach
:
- Bal.:1 KIT
- Popis:Hydrazine color disc test kit, HY-2
- Testů v kitu:300
- Rozsah:0 to 1 mg/l
Proper maintenance and economy of operation of modern boilers and cooling towers require a careful balance between water quality parameters and treatment chemicals. This kit measures hydrazine, a common oxygen scavenger in boiler water, using the simple color disc method.
: Includes two 100 ml bottles of deionised water, one reagent, two stoppers, one colour disc comparator, one colour disc, instructions, and a carrying case.