Orders LinkContactUsLinkComponent
Test kits, COD cell test, Spectroquant®, Supelco®
 :   Merck
Test kits, COD cell test, Spectroquant®, Supelco®
 :  Merck
Test kits, COD cell test, Spectroquant®, Supelco®
Test kits, COD cell test, Spectroquant®, Supelco®
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and wastewater
*8 COD cell tests fully comply with ISO 15705.
  • Catalog No:
  • 1.14560.0001
  • 1.01796.0001
  • 1.14540.0001
  • 1.14895.0001
  • 1.14541.0001
  • 1.14690.0001
  • 1.14691.0001
  • 1.14555.0001
  • 1.01797.0001
  • 1.09772.0001
  • 1.09773.0001
  • M-Clarity quality level:
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
  • MQ100
Product Family Options
  • Sekvence
    Popis
    Rozsah
    Chemická metoda
    Pro jednotku
    Bal.
  • 20
    Kyvetový test COD*⁷⁻¹
    4,0−40,0 mg/L
    Chromosulfurická oxidace
    Prove, Nova 30/60
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 630,00 Kč
    4 630,00 Kč
    1 * 25 Tests
       4 630,00 Kč
    Specifications
    Cat. No.
    1.14560.0001
    M-Clarity quality level
    MQ100
  • 22
    Kyvetový test COD*⁸
    5,0−80,0 mg/L
    Chromosulfurická oxidace
    Prove, Nova 30/60, Multy, Move
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 630,00 Kč
    4 630,00 Kč
    1 * 25 Tests
       4 630,00 Kč
    Specifications
    Cat. No.
    1.01796.0001
    M-Clarity quality level
  • 23
    Kyvetový test COD*⁷⁻¹
    10−150 mg/L
    Chromosulfurická oxidace
    Prove, Nova 30/60, Multy, Move, Picco
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 630,00 Kč
    4 630,00 Kč
    1 * 25 Tests
       4 630,00 Kč
    Specifications
    Cat. No.
    1.14540.0001
    M-Clarity quality level
    MQ100
  • 24
    Kyvetový test COD*⁷⁻¹
    15−300 mg/L
    Chromosulfurická oxidace
    Prove, Nova 30/60, Multy, Move
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 630,00 Kč
    4 630,00 Kč
    1 * 25 Tests
       4 630,00 Kč
    Specifications
    Cat. No.
    1.14895.0001
    M-Clarity quality level
    MQ100
  • 25
    Kyvetový test COD*⁷⁻¹
    25−1 500 mg/L
    Chromosulfurická oxidace
    Prove, Nova 30/60, Multy, Move, Picco
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 630,00 Kč
    4 630,00 Kč
    1 * 25 Tests
       4 630,00 Kč
    Specifications
    Cat. No.
    1.14541.0001
    M-Clarity quality level
    MQ100
  • 26
    Kyvetový test COD*⁷⁻¹
    50−500 mg/L
    Chromosulfurická oxidace
    Prove, Nova 30/60, Multy, Move
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 630,00 Kč
    4 630,00 Kč
    1 * 25 Tests
       4 630,00 Kč
    Specifications
    Cat. No.
    1.14690.0001
    M-Clarity quality level
    MQ100
  • 27
    Kyvetový test COD*⁷⁻¹
    300−3 500 mg/L
    Chromosulfurická oxidace
    Prove, Nova 30/60, Multy, Move
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 630,00 Kč
    4 630,00 Kč
    1 * 25 Tests
       4 630,00 Kč
    Specifications
    Cat. No.
    1.14691.0001
    M-Clarity quality level
    MQ100
  • 28
    Kyvetový test COD*⁷⁻¹
    500−10 000 mg/l
    Chromosulfurická oxidace
    Prove, Nova 30/60, Multy, Move, Picco
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 630,00 Kč
    4 630,00 Kč
    1 * 25 Tests
       4 630,00 Kč
    Specifications
    Cat. No.
    1.14555.0001
    M-Clarity quality level
    MQ100
  • 29
    Kyvetový test COD*⁸
    5000−90000 mg/L
    Chromosulfurická oxidace
    Prove, Nova 30/60, Multy, Move
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 620,00 Kč
    4 620,00 Kč
    1 * 25 Tests
       4 620,00 Kč
    Specifications
    Cat. No.
    1.01797.0001
    M-Clarity quality level
  • 32
    Kyvetový test COD (bez Hg)
    10−150 mg/L
    Chromosulfurická oxidace
    Prove, Nova 30/60, Move, Multy
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 3 860,00 Kč
    3 860,00 Kč
    1 * 25 Tests
       3 860,00 Kč
    Specifications
    Cat. No.
    1.09772.0001
    M-Clarity quality level
    MQ100
  • 33
    Kyvetový test COD (bez Hg)
    100−1500 mg/L
    Chromosulfurická oxidace
    Prove, Nova 30/60, Move, Multy
    25 Tests
    1 * 25 Tests
    1 * 25 Tests - 4 620,00 Kč
    4 620,00 Kč
    1 * 25 Tests
       4 620,00 Kč
    Specifications
    Cat. No.
    1.09773.0001
    M-Clarity quality level
    MQ100
COD Cell Test for seawater / high chloride contents
  •  
    Pro jednotku
    Chemická metoda
    Rozsah
    Popis
    Bal.
    Sekvence
  • Prove, Nova 30/60, Multy, Move
    Chromosulphuric oxidation
    5,0 - 60,0 mg/L
    COD cell test for seawater
    25 Tests
    30
    1 * 25 Tests
    1 * 25 Tests - 6 720,00 Kč
    6 720,00 Kč
    1 * 25 Tests
       6 720,00 Kč
  •  
    Pro jednotku
    Chemická metoda
    Rozsah
    Popis
    Bal.
    Sekvence
  • Prove, Nova 30/60, Multy, Move
    Chromosulphuric oxidation
    50 - 3000 mg/L
    COD cell test for seawater
    25 Tests
    31
    1 * 25 Tests
    1 * 25 Tests - 6 720,00 Kč
    6 720,00 Kč
    1 * 25 Tests
       6 720,00 Kč
Accessories
  •  
    Sekvence
    Popis
    Bal.
  • 1.15955.0001
    02
    Nástavce dávkovací k CHSK analýze mořské vody 1.17059.0001
    1
    1 * 1 KS - 6 790,00 Kč
  •  
    Sekvence
    Typ balení
    Bal.
  • 1.06733.0501
    3645
    Plastová láhev
    500 g
    1 * 500 g - 2 780,00 Kč