NMR tubes, economy, Suprasil® EPR tubes, SP Wilmad-LabGlass
Katalogové číslo WILM724-SQ-100M
Dodavatel: SP Industries
Specifikace
- Délka:100 mm
- Bal.:1
Specifikace
O tomto produktu
Tubes made of synthetic material suppress major impurities during EPR.
Delivery: Each EPR tube comes with a disposable grade cap.