Surgical gloves, polyisoprene, sterile, Gammex® Non-Latex PI Micro
These thin gloves made from synthetic polyisoprene offer an optimal level of sensitivity, comfort, and protection against natural rubber latex allergies.
- Powder-free, micro-textured finish
- DPG-free polyisoprene option for healthcare workers suffering from latex allergies and chemical allergies
- White colour makes it ideal as a double-gloving solution, together with the green GAMMEX® Non-Latex PI underglove
- Beaded cuffs with adhesive band
- External surface: Micro-textured and chlorinated and siliconised; internal surface: PU coated and siliconised
Ideal for delicate procedures in ophthalmic, plastic, neurologic, cardiovascular, and reconstructive surgeries.
: EN 455 (1-4), EN ISO 374-1 Type B (KMPT), EN ISO 374-5 VIRUS, EN 16523-1, Cat. III; AQL 0,65
- Catalog No:
- ANSM340057055
- ANSM340057060
- ANSM340057070
- ANSM340057065
- ANSM340057080
- ANSM340057085
- ANSM340057075
- ANSM340057090
- Model:
- Non-Latex PI Micro
- Non-Latex PI Micro
- Non-Latex PI Micro
- Non-Latex PI Micro
- Non-Latex PI Micro
- Non-Latex PI Micro
- Non-Latex PI Micro
- Non-Latex PI Micro
- Obouruké/jednoruké:
- Jednoruké
- Jednoruké
- Jednoruké
- Jednoruké
- Jednoruké
- Jednoruké
- Jednoruké
- Jednoruké
- Jednorázové/znovupoužitelné:
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Jednorázové
- Latex/bez latexu:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Pudrované/nepudrované:
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Nepudrované
- Provedení:
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Micro-textured
- Typ manžety:
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Rolovaná
- Norma EN:
- EN 455 (1-4), EN ISO 374-1 Type B (KMPT), EN ISO 374-5 VIRUS, EN 16523-1
- EN 455 (1-4), EN ISO 374-1 Type B (KMPT), EN ISO 374-5 VIRUS, EN 16523-1
- EN 455 (1-4), EN ISO 374-1 Type B (KMPT), EN ISO 374-5 VIRUS, EN 16523-1
- EN 455 (1-4), EN ISO 374-1 Type B (KMPT), EN ISO 374-5 VIRUS, EN 16523-1
- EN 455 (1-4), EN ISO 374-1 Type B (KMPT), EN ISO 374-5 VIRUS, EN 16523-1
- EN 455 (1-4), EN ISO 374-1 Type B (KMPT), EN ISO 374-5 VIRUS, EN 16523-1
- EN 455 (1-4), EN ISO 374-1 Type B (KMPT), EN ISO 374-5 VIRUS, EN 16523-1
- EN 455 (1-4), EN ISO 374-1 Type B (KMPT), EN ISO 374-5 VIRUS, EN 16523-1
Frequently Bought Together