Adapter kit
Adaptor kit (4 mm male / 4 mm female)
Katalogové číslo PSA410
Dodavatel: Cytiva
Přejít na:
Product Details & Documents
Connects 4 mm output jack to 4 mm high voltage leads.
Note that this adaptor kit is not CE compliant.
Informace pro objednávání: Supplied with Allen wrench.
Documents
Cytiva CyDye Licensing Statements Cytiva General Licensing Statements
Další podrobnosti o produktu
Specifikace
- Popis:Adaptor kit (4 mm male / 4 mm female)