Standards for spectrophotometer calibration, Supelco®
Katalogové číslo 1.08159.0100
Supplier: Merck
CAS Number:
Specifications
- Bal.:100 mL
- Popis:Holmium perchlorate, density 1,136 g/cm3, soluble at 20 °C, with absorbance of 241,1±0,4 nm
- M-Clarity quality level:MQ100
Specifications
About this item
Standard solutions are used for the calibration of spectrophotometers.