Hydroxynaphthol blue indicator for metal titration, Supelco®
Catalog # 1.04593.0025
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle
- Bal.:25 g
- Sekvence:3308
Specifications
Specification Test Results
- Appearance of solution:conforms
- Loss on Drying (105 °C):≤ 5 %
- Suitability as indicator (for metal titration):conforms
Specification Test Results
About this item
:
:620.47 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:264-197-7
CAS Number:63451-35-4
Formula:C₂₀H₁₁N₂Na₃O₁₁S₃
MDL Number:MFCD00036393