Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-PTGES3 Rabbit Polyclonal Antibody
  BOSSBS-1909R
 :  Bioss
Anti-PTGES3 Rabbit Polyclonal Antibody
  BOSSBS-1909R
 :  Bioss

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Prostaglandin E Synthase 3
  • Symbol antigenu:
    PTGES3
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Formulář:
    liquid
  • Identifikační číslo genu:
    10728
  • Synonyma antigenu:
    Co chaperone p23|Progesterone receptor complex|Sid 3177|Telomerase binding protein p23|TEBP|Hsp90 co chaperone
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Koncentrace:
    1 ug/ul
  • Transportní teplota:
    4 °C
  • Čištění:
    Purified by Protein A.
  • Rozměry protilátky:
    100 μg
  • Bal.:
    100 µl

 

 

Molecular chaperone that localizes to genomic response elements in a hormone-dependent manner and disrupts receptor-mediated transcriptional activation, by promoting disassembly of transcriptional regulatory complexes. Belongs to the p23/wos2 family.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: PTGES3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat