Order Entry
Czech Republic
ContactUsLinkComponent
Anti-LAMTOR3 Rabbit Polyclonal Antibody
Anti-LAMTOR3 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13688R
Supplier:  Bioss
Anti-LAMTOR3 Rabbit Polyclonal Antibody
Catalog # BOSSBS-13688R
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Late Endosomal/Lysosomal Adaptor, MAPK And MTOR Activator 3
  • Symbol antigenu:
    LAMTOR3
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Formulář:
    liquid
  • Synonyma antigenu:
    MEK-binding partner 1|Mitogen activated protein kinase kinase 1 interacting protein 1|Ragulator complex protein LAMTOR3.|Mitogen-activated protein kinase kinase 1-interacting protein 1|MP1|MEK partner 1|MEK binding partner 1|LTOR3_HUMAN|Mitogen-activated protein kinase scaffold protein 1
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Koncentrace:
    1 ug/ul
  • Transportní teplota:
    4 °C
  • Čištění:
    Purified by Protein A.
  • Rozměry protilátky:
    100 μg
  • Bal.:
    100 µl

Specifications

About this item

This gene encodes a scaffold protein that functions in the extracellular signal-regulated kinase (ERK) cascade. The protein is localized to late endosomes by the mitogen-activated protein-binding protein-interacting protein, and binds specifically to MAP kinase kinase MAP2K1/MEK1, MAP kinase MAPK3/ERK1, and MAP kinase MAPK1/ERK2. Studies of the orthologous gene in mouse indicate that it regulates late endosomal traffic and cell proliferation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene. A pseudogene of this gene is located on the long arm of chromosome 13. [provided by RefSeq, Aug 2011]

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: LAMTOR3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat