Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Blood Group Lewis a Rabbit Polyclonal Antibody
Anti-Blood Group Lewis a Rabbit Polyclonal Antibody
Catalog # BOSSBS-12870R
Supplier:  Bioss
Anti-Blood Group Lewis a Rabbit Polyclonal Antibody
Catalog # BOSSBS-12870R
Supplier:  Bioss

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Blood Group Lewis a
  • Symbol antigenu:
    Blood Group Lewis a
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Izotyp:
    IgG
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Formulář:
    liquid
  • Synonyma antigenu:
    Lewis A antigen.
  • Skladovací pufr:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0,09% sodium azide. Store at -20°C for 12 months.
  • Koncentrace:
    1 ug/ul
  • Transportní teplota:
    4 °C
  • Čištění:
    Purified by Protein A.
  • Rozměry protilátky:
    100 μg
  • Bal.:
    100 µl

Specifications

About this item

Glycosyltransferases that mediate the regio- and stereoselective transfer of sugars, such as the fucosyltransferases, determine cell surface-carbohydrate profiles, which is an essential interface for biological recognition processes. Fucosyltransferases catalyze the covalent association of fucose to different positional linkages in sugar acceptor molecules. The carbohydrate moieties generated and covalently attached to cell surfaces are necessary to ensure a surface contour that satisfies physiological roles, which are reliant on adhesion molecules such as Selectins (1-3). Hematopoietic lineages rely on Fucosyltransferases to confer a surface carbohydrate phenotype, which mediates proper cell adhesion molecule recruitment and cell trafficking (4-6). Blood Group Lewis a is a carbohydrate determinant carried on both glycolipids and glycoproteins.

WB(1:100-1000), IHC-P(1:100-500), IF(IHC-P)(1:50-200)

Type: Primary
Antigen: Blood Group Lewis a
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human