Schlenk sample tube, SP Wilmad-LabGlass
Katalogové číslo WILMLG-10800-100
Dodavatel: SP Industries
Specifikace
- Kapacita:5 ml
- Bal.:1
Specifikace
O tomto produktu
Tube incorporates a grease-free high vacuum 0 to 4 mm valve and is suited for pressure/vacuum applications. Ideal for reaction and storage of air-sensitive compounds. Sidearm can be connected to inert atmosphere/vacuum source or a manifold.