Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 Kč
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Czech Republic
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Záznamníky dat (Dataloggery)
Accessories for temperature and humidity data logger AiroSensor
Accessories for temperature and humidity data logger AiroSensor
Accessories for temperature and humidity data logger AiroSensor
Catalog # SENA0630125977934
Supplier:
SenseAnywhere
Expand image
Accessories for temperature and humidity data logger AiroSensor
Catalog # SENA0630125977934
Supplier:
SenseAnywhere
Specifications
Popis:
SwitchHolder
Hmotnost:
14 g
Š×H×V:
52×25×34 mm
Bal.:
1
Specifications
About this item
Product Details
Product Details
Prepaid subscription card with activation code
Product Information