


A-Phellandrene for synthesis, Sigma-Aldrich®
8.14893.0100
: Merck
:
- Typ balení:Glass bottle
- Bal.:100 ml
- Sekvence:2440
- Solubility:2.86 mg/l (25 °C)
- Molar Mass:136.23 g/mol
- Boiling Point:175 °C (1013 hPa)
- Flash Point:47 °C
- Density:0.84 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Storage temperature:+15...+25 °C
- Assay (GC, area%):≥60,0 Area%
:
:136.238 g/mol
:3,III
:M-Clarity™ quality level = MQ200
:202-792-5
:1993
:GHS02
:GHS08
:GHS07
:Ambient
:99-83-2
:C₁₀H₁₆
:MFCD00040419