ContactUsLinkComponent
Biocide, sterile, filtered, Klercide™ Quat | Biguanide
Biocide, sterile, filtered, Klercide™ Quat | Biguanide
  141-1155
Biocide, sterile, filtered, Klercide™ Quat | Biguanide
  141-1155
  • Popis:
    Klercide Sterile Quat | Biguanide spray
  • Sekvence:
    01
  • Balené:
    1 litre bottle with trigger spray
  • Bal.:
    6
Blend of a quaternary ammonium compound and a biguanide with broad spectrum activity. Suitable for ISO Class 5; Grade A and B (microbial count) areas. The biocide blend is 0,2 micron filtered, filled and double bagged in a Grade C cleanroom before gamma irradiation to a validated procedure.
  • CEN and isolate tested
  • Safe and effective: non toxic and non corrosive
  • Fully compatible and ideal for a rotational regime
  • SteriShield Delivery System™ ensures 6 months in-use shelf life
 : EN standards: EN 1276 - Bactericidal activity; EN 1650 - Fungicidal activity; EN 13697 Bactericidal surface test.
 : Use biocides safely. Always read the label and product information before use.