ContactUsLinkComponent
Reagents for non Merck photometers, Spectroquant®, Supelco®
Katalogové číslo 1.19254.0001
Supplier:  Merck
CAS Number:  
undefined
Reagents for non Merck photometers, Spectroquant®, Supelco®
Katalogové číslo 1.19254.0001
Supplier:  Merck
CAS Number:  
Specifications
  • Bal.:
    100 Tests
  • Popis:
    Chlorine powder packs (10 ml free chlorine samples)
  • Sekvence:
    01
  • Testů v kitu:
    100
  • Rozsah:
    0 - 2,00 mg/l
Specifications
About this item
Spectroquant® test kits are suitable for use only in conjunction with Hach instruments. They do not require any further device-specific calibration.