ContactUsLinkComponent
HPLC columns, Hibar®, LiChrospher® 100 CN
  1.50182.7071
 :  Merck
undefined
HPLC columns, Hibar®, LiChrospher® 100 CN
  1.50182.7071
 :  Merck
  • Formát kolony:
    Analytical
  • Ø vnitř.:
    4,00 mm
  • Velikost částic:
    5 µm
  • Velikost pórů:
    100 Å
  • Délka:
    250 mm
  • Sekvence:
    03
  • Fáze:
    CN
  • Bal.:
    1
Stainless steel Hibar® columns are complete with endfittings. These columns are available with various types of LiChrospher® sorbents.
When using a guard column with a Hibar® RT column, we recommend part number 1.51487.0001 guard column cartridge holder for 4×4 mm guard column cartridges LiChroCART®.