Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-XRCC4 Polyclonal Antibody Pair
  ABNOH00007518-AP21
 :  Abnova
Anti-XRCC4 Polyclonal Antibody Pair
  ABNOH00007518-AP21
 :  Abnova

 

  • Název antigenu:
    X-ray Repair Complementing Defective Repair in Chinese Hamster cells 4
  • Symbol antigenu:
    XRCC4
  • Klonovalita:
    Polyclonal
  • Hostitel:
    Multiple
  • Reaktivita:
    Human
  • Identifikační číslo genu:
    7518
  • Synonyma antigenu:
    SSMED
  • Skladovací teplota:
    Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
  • Rozměry protilátky:
    1 Set
  • Bal.:
    1 SET

 

 

This Set comes with matched Antibody Pair to detect and quantify protein level of human XRCC4.

Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-XRCC4 (100 µg); 2. Detection Antibody: Mouse purified Polyclonal Anti-XRCC4 (20 µg);

Type:
Antigen: XRCC4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human