Anti-ID2H Polyclonal Antibody Pair
Katalogové číslo ABNOH00003398-AP21
Dodavatel: Abnova
Specifikace
- Název antigenu:Inhibitor Of Differentiation 2
- Symbol antigenu:ID2H
- Klonovalita:Polyclonal
- Hostitel:Multiple
- Reaktivita:Human
- Identifikační číslo genu:3398
- Synonyma antigenu:GIG8|ID2A|Cell Growth-Inhibiting Gene 8|Inhibitor Of DNA Binding 2|bHLHb26|Dominant Negative Helix-Loop-Helix Protein|ID2H|DNA-Binding Protein Inhibitor ID2
- Skladovací teplota:Store reagents of the antibody pair set at –20 °C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to –20 °C storage immediately after use.
- Rozměry protilátky:1 Set
- Bal.:1 SET
Specifikace
O tomto produktu
This Set comes with matched Antibody Pair to detect and quantify protein level of human ID2.
Antibody pair set content: 1. Capture Antibody: Rabbit MaxPab® affinity purified Polyclonal Anti-ID2 (100 µg); 2. Detection Antibody: Mouse Polyclonal Anti-ID2 (40 µl);
Type:
Antigen: ID2H
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity: Human