Hydrophobic interaction chromatography column, HiTrap™ Capto™ Phenyl ImpRes
Catalog # GEHE17-5484-12
Supplier: Cytiva
Specifications
- Velikost částic:36 - 44 µm
- Maximální tlak:5 bar
- Kapacita:5 ml
- Popis:HiTrap™ Capto™ Phenyl ImpRes
- Sekvence:02
- Rozsah pH:2 - 13
- Hardware kolony:PP
- Rozměry lůžka:16×25 mm
- Bal.:5
Specifications
About this item
HiTrap Capto Phenyl ImpRes is prepacked with Capto Phenyl ImpRes media for screening and small-scale protein purifications using hydrophobic interaction chromatography.
- High-throughput medium for intermediate and polishing protein purification, excellent choice for process-scale purification
- Columns for screening of selectivity, binding and elution conditions, as well as small scale purifications
- Excellent chemical stability allows cleaning with all standard CIP solutions, including 1 M sodium hydroxide
The combined properties of high binding capacity, high flow rate, and low back pressure reduces process cycle times and improves productivity at process scale.