Solvent filter/degasser
03, RC filter membrane, 0,45 μm, 47 mm
Katalogové číslo HEWL3150-0576
Dodavatel: Agilent
Přejít na:
Product Details & Documents
It prevents the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph.
- Degasses eluents as particulates are removed
- Increases solvent inlet lifetime
- Eliminates pump downtime caused by air locks and particulates in check valves
- Decreases piston wear, while increasing column life
Specifikace
- Ø:47 mm
- Velikost pórů:0,45 μm
- Popis:RC filter membrane
- Sekvence:03