Anti-TFR2 Mouse Monoclonal Antibody [clone: 3C5]
Catalog # ABNOH00007036-M01
Supplier: Abnova
Specifications
- Druh protilátky:Primární
- Název antigenu:transferrin receptor 2
- Symbol antigenu:TFR2
- Klonovalita:Monoclonal
- Klon:3C5
- Spojení:Unconjugated
- ELISA:Yes
- Hostitel:Mouse
- Izotyp:IgG1 kappa
- Reaktivita:Human
- Western blot:Yes
- Křížová adsorbce:No
- Formát:Liquid
- Identifikační číslo genu:7036
- Synonyma antigenu:HFE3|TFRC2
- Počet aminokyselin:631 to 731
- Skladovací pufr:1x PBS, pH 7,4
- Sekvence:IRLSHDRLLPLDFGRYGDVVLRHIGNLNEFSGDLKARGLTLQWVYSARGDYIRAAEKLRQEIYSSEERDERLTRMYNVRIMRVEFYFLSQYVSPADSPFR*
- Skladovací teplota:Store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Imunogen:TFR2 (NP_003218, 631 a.a. ~ 731 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
- Rozměry protilátky:100 µg
- Bal.:100 µg
Specifications
About this item
Mouse monoclonal antibody raised against a partial recombinant TFR2.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: TFR2
Clonality: Monoclonal
Clone: 3C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human