Anti-MAP2K4 Rabbit Polyclonal Antibody
Catalog # ABNOPAB5570
Supplier: Abnova
Specifications
- Druh protilátky:Primární
- Název antigenu:Mitogen-activated protein kinase Kinase 4
- Symbol antigenu:MAP2K4
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Reaktivita:Human,Rat,Mouse
- Western blot:Yes
- Křížová adsorbce:No
- Formát:Liquid
- Identifikační číslo genu:6416
- Synonyma antigenu:MKK4|SAPKK-1|PRKMK4|SAPKK1|MAPKK4|SERK1|SKK1|MEK4|JNKK1|JNKK|SEK1
- Skladovací pufr:PBS, 150 mM NaCl, pH 7,4 (50% glycerol, 0,02% sodium azide)
- Skladovací teplota:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Imunogen:A synthetic peptide corresponding to residues surrounding S80 of human MAP2K4.
- Rozměry protilátky:100 μg
- Bal.:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of MAP2K4.
Recommended Dilutions: Western Blot: 1:500-1:1000
Type: Primary
Antigen: MAP2K4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat