Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC25A20 Rabbit Polyclonal Antibody
Anti-SLC25A20 Rabbit Polyclonal Antibody
Catalog # ABGEAP19135B
Supplier:  Abgent
Anti-SLC25A20 Rabbit Polyclonal Antibody
Catalog # ABGEAP19135B
Supplier:  Abgent

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    solute carrier family 25 (carnitine/acylcarnitine translocase), member 20
  • Symbol antigenu:
    Solute carrier family 25, member 20
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Reaktivita:
    Mouse
  • Western blot:
    Yes
  • Epitop:
    C-terminal
  • Identifikační číslo genu:
    788
  • Synonyma antigenu:
    CACT|CAC
  • Počet aminokyselin:
    235 to 264
  • Skladovací pufr:
    pH 7,4
  • Molekulární hmotnost:
    32.944 kDa
  • Sekvence:
    RDVLRELIRDEGVTSC
  • Skladovací teplota:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Koncentrace:
    1.320
  • Transportní teplota:
    4 °C
  • Imunogen:
    This SLC25A20 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 235-264 amino acids from the C-terminal region of human SLC25A20.
  • Čištění:
    Protein A & Peptide affinity purification
  • Bal.:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SLC25A20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse