Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Methylated Lysine Rabbit Polyclonal Antibody (Biotin)
Anti-Methylated Lysine Rabbit Polyclonal Antibody (Biotin)
  CAYM13728-400
Anti-Methylated Lysine Rabbit Polyclonal Antibody (Biotin)
  CAYM13728-400

 

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    Methylated Lysine
  • Klonovalita:
    Polyclonal
  • Spojení:
    Biotin
  • ELISA:
    Yes
  • Imunoprecipitace:
    Yes
  • Western blot:
    Yes
  • Křížová adsorbce:
    No
  • Skladovací pufr:
    PBS, pH 7,0, containing 50% glycerol and 0,01% sodium azide
  • Skladovací teplota:
    −20 °C
  • Transportní teplota:
    −20 °C
  • Čištění:
    purified by affinity chromatography
  • Bal.:
    400 µl

 

 

Type: Primary
Antigen: Methylated Lysine
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: multi-species