Anti-S1P5 Rabbit Polyclonal Antibody
Catalog # CAYM13490-1
Supplier: Cayman Chemical
Specifications
- Druh protilátky:Primární
- Symbol antigenu:S1P5
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Imunochemie:Yes
- Izotyp:IgG
- Reaktivita:Monkey,Human,Dog,Rat,Mouse
- Western blot:Yes
- Křížová adsorbce:No
- Skladovací pufr:PBS containing 0,05% BSA and 0,05% sodium azide
- Skladovací teplota:–20 °C
- Transportní teplota:–20 °C
- Imunogen:peptide from human S1P5 within the region of amino acids 1-50
- Čištění:purified by Protein G chromatography
- Rozměry protilátky:1 Ea
- Bal.:1
Specifications
About this item
Type: Primary
Antigen: S1P5
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Dog, Human, Monkey, Mouse, Rat