Anti-GluR1 Rabbit Polyclonal Antibody
Catalog # CAYM10602-1
Supplier: Cayman Chemical
Specifications
- Druh protilátky:Primární
- Symbol antigenu:GLUR1
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Rat
- Western blot:Yes
- Křížová adsorbce:No
- Modifikace:phospho-Ser831
- Skladovací teplota:–20 °C
- Transportní teplota:–20 °C
- Imunogen:phosphopeptide corresponding to amino acid residues surrounding the phospho-Ser831 of GluR1
- Čištění:purified by affinity chromatography
- Rozměry protilátky:1 Ea
- Bal.:1
Specifications
About this item
Type: Primary
Antigen: GluR1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat