Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IgG (H+L) Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Anti-IgG (H+L) Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # ROCK704-408-002
Anti-IgG (H+L) Rabbit Polyclonal Antibody (RPE (R-Phycoerythrin))
Catalog # ROCK704-408-002

Specifications

  • Druh protilátky:
    Sekundární
  • Symbol antigenu:
    IgG (H+L)
  • Klonovalita:
    Polyclonal
  • Spojení:
    RPE (R-Phycoerythrin)
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Rabbit
  • ImunoFluorescence:
    Yes
  • Reaktivita:
    Dog
  • Křížová adsorbce:
    No
  • Formát:
    Antibody fraction
  • Formulář:
    Lyophilized
  • Fragment protilátky:
    F(ab')2
  • Skladovací pufr:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Skladovací teplota:
    +4 °C
  • Transportní teplota:
    Ambient
  • Imunogen:
    Dog IgG whole molecule
  • Čištění:
    purified by immunoaffinity chromatography
  • Rozměry protilátky:
    500 µg
  • Bal.:
    500 µG

Specifications

About this item

Recommended Dilutions: Immunofluorescence: 1:100 - 1:250; Flow Cytometry: 1:100 - 1:250
Restore with deionized water (or equivalent); 1.0 ml
Do not freeze! Light sensitive!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype:
Reactivity: Dog