Anti-GST Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # ROCK600-400-200
Supplier: Rockland Immunochemicals
Specifications
- Druh protilátky:Primární
- Název antigenu:Glutathione-S-Transferase
- Symbol antigenu:GST
- Klonovalita:Polyclonal
- Spojení:TRITC (Tetramethylrhodamine Isothiocyanate)
- Průtoková cytometrie:Yes
- Hostitel:Rabbit
- ImunoFluorescence:Yes
- Reaktivita:Mouse
- ChIP:Yes
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Rabbit
Isotype:
Reactivity: