Anti-AKT1 Mouse Polyclonal Antibody (Unconjugated) [clone: 14E5.16C8.25F6]
Catalog # ROCK200-301-401
Supplier: Rockland Immunochemicals
Specifications
- Druh protilátky:Primární
- Název antigenu:v-akt murine thymoma viral oncogene homolog 1
- Symbol antigenu:AKT1
- Klonovalita:Polyclonal
- Klon:14E5.16C8.25F6
- Spojení:Unconjugated
- ELISA:Yes
- Průtoková cytometrie:Yes
- Hostitel:Mouse
- Imunochemie:Yes
- Imunoprecipitace:Yes
- Izotyp:IgG1 kappa
- Reaktivita:Chimpanzee,Human,Rat,Mouse
- Western blot:Yes
- Epitop:Internal
- Křížová adsorbce:No
- Formát:Antibody fraction
- Formulář:Liquid (sterile filtered)
- Identifikační číslo genu:207
- Synonyma antigenu:CWS6|RAC|RAC-ALPHA|PRKBA|PKB|AKT|PKB-ALPHA
- Skladovací pufr:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Skladovací teplota:–20 °C
- Transportní teplota:Dry Ice
- Imunogen:Anti-AKT Antibody was produced by repeated immunizations with a synthetic peptide corresponding to residues internal to the human AKT1 Protein.
- Čištění:Protein A purified
- Rozměry protilátky:100 µg
- Bal.:100 µG
Specifications
About this item
Anti-AKT Antibody is suitable for ELISA, immunochemistry, immunoprecipitation and western blotting. Expect a band approximately 54 - 56 kDa in size corresponding to AKT Protein by western blotting in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:500 - 1:3000; Immunochemistry: 20 µg/ml;
Type: Primary
Antigen: AKT1
Clonality: Polyclonal
Clone: 14E5.16C8.25F6
Conjugation: Unconjugated
Epitope: Center
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat, Chimpanzee