Orders LinkContactUsLinkComponent
Attachable screw thread tubes
Attachable screw thread tubes
SVL 15, 16 mm, 110 mm
Catalog # WITG0.182.315
Supplier:   witeg Labortechnik
Attachable screw thread tubes
Catalog # WITG0.182.315
Supplier:  witeg Labortechnik
Attachable screw thread tubes
Attachable screw thread tubes
SVL 15, 16 mm, 110 mm
Price
56,40 Kč
Log in for availability
1 * 1 KS
Product Details & Documents
DURAN®, borosilicate glass 3.3, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
DIN screw thread SVL.
Specifications
  • Závit:
    SVL 15
  • Vnějš. Ø zkumavky:
    16 mm
  • Celková délka:
    110 mm