ContactUsLinkComponent
K Probe for 735 (Testo)
K Probe for 735 (Testo)
Catalog # 620-1194
Supplier:  Testo
undefined
K Probe for 735 (Testo)
Catalog # 620-1194
Supplier:  Testo
Specifications
  • Rozsah:
    −60…+250 °C
  • Popis:
    Waterproof fast needle probe, K
  • Sekvence:
    11
  • Bal.:
    1