Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Csnk2a1 Rabbit Polyclonal Antibody
Anti-Csnk2a1 Rabbit Polyclonal Antibody
Catalog # ORIGTA328199
Supplier:  OriGene
Anti-Csnk2a1 Rabbit Polyclonal Antibody
Catalog # ORIGTA328199
Supplier:  OriGene

Specifications

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    Csnk2a1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Reaktivita:
    Drosophila,
    Rabbit,
    Xenopus,
    Chicken,
    C. elegans,
    Human,
    Bovine,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    near N-terminus
  • Křížová adsorbce:
    No
  • Formulář:
    Liquid
  • Počet aminokyselin:
    28 to 50
  • Skladovací pufr:
    PBS with 0,09% (W/V) sodium azide
  • Skladovací teplota:
    –20 °C
  • Transportní teplota:
    –20 °C
  • Imunogen:
    This Rat Csnk2a1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 28-50 amino acids from the N-terminal region of Rat Csnk2a1.
  • Čištění:
    purified by Protein A chromatography, followed by affinity purification
  • Rozměry protilátky:
    400 μl
  • Bal.:
    400 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: Csnk2a1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near N-terminus
Host: Rabbit
Isotype:
Reactivity: Bovine, C.elegans, Chicken, Drosophila, Human, Mouse, Rabbit, Rat, Xenopus