Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Scn2a1 Rabbit Polyclonal Antibody
Anti-Scn2a1 Rabbit Polyclonal Antibody
  ORIGTA329028
 :  OriGene
Anti-Scn2a1 Rabbit Polyclonal Antibody
  ORIGTA329028
 :  OriGene

 

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    Scn2a1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Imunoprecipitace:
    Yes
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Křížová adsorbce:
    No
  • Formulář:
    Lyophilized
  • Počet aminokyselin:
    467 to 485
  • Skladovací pufr:
    Lyophilized. Concentration before lyophilization ~0,8mg/ml (lot dependent, please refer to CoA along with shipment for actual concentration). Buffer before lyophilization: phosphate buffered saline (PBS), pH 7,4, 1% BSA, 0,05% NaN3.
  • Sekvence:
    C-ASAESRDFSGAGGIGVFSE
  • Skladovací teplota:
    –20 °C
  • Transportní teplota:
    –20 °C
  • Imunogen:
    Peptide (C)ASAESRDFSGAGGIGVFSE, corresponding to amino acid residues 467-485 of rat Nav1.2.? ? Intracellular loop between domains I and II .
  • Čištění:
    purified by antigen affinity chromatography
  • Rozměry protilátky:
    200 μl
  • Bal.:
    200 µl

 

 

Recommended Dilutions: Western Blot: 1:200-1:2000; Immunochemistry: 1:100-1:3000

Type: Primary
Antigen: Scn2a1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat