Order Entry
Czech Republic
ContactUsLinkComponent
Anti-ASIC2 Rabbit Polyclonal Antibody
Anti-ASIC2 Rabbit Polyclonal Antibody
Catalog # ORIGTA329036
Supplier:  OriGene
Anti-ASIC2 Rabbit Polyclonal Antibody
Catalog # ORIGTA329036
Supplier:  OriGene

Specifications

  • Druh protilátky:
    Primární
  • Název antigenu:
    Acid sensing (proton-gated) ion channel 2
  • Symbol antigenu:
    ASIC2
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Reaktivita:
    Rat
  • Western blot:
    Yes
  • Křížová adsorbce:
    No
  • Formulář:
    Lyophilized
  • Počet aminokyselin:
    2 to 18
  • Skladovací pufr:
    Lyophilized. Concentration before lyophilization ~0,8mg/ml (lot dependent, please refer to CoA along with shipment for actual concentration). Buffer before lyophilization: Phosphate buffered saline (PBS), pH 7,4, 1% BSA, 5% sucrose, 0,025% NaN3.
  • Sekvence:
    DLKESPSEGSLQPSSIQC
  • Skladovací teplota:
    –20 °C
  • Transportní teplota:
    –20 °C
  • Imunogen:
    Peptide DLKESPSEGSLQPSSIQC, corresponding to amino acid residues 2-18 of human ASIC2a. Intracellular, N-terminus.
  • Čištění:
    purified by antigen affinity chromatography
  • Rozměry protilátky:
    200 μl
  • Bal.:
    200 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1:200-1:2000

Type: Primary
Antigen: ASIC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Rat