Anti-DTYMK Mouse Monoclonal Antibody [clone: 1D2]
ORIGCF503497
: OriGene
- Druh protilátky:Primární
- Název antigenu:Deoxythymidylate kinase
- Symbol antigenu:DTYMK
- Klonovalita:Monoclonal
- Klon:1D2
- Spojení:Unconjugated
- Hostitel:Mouse
- ImunoFluorescence:Yes
- Izotyp:IgG1
- Reaktivita:Human
- Western blot:Yes
- Epitop:Full length
- Křížová adsorbce:No
- Formulář:Lyophilized
- Skladovací pufr:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molekulární hmotnost:23.6 kDa
- Skladovací teplota:–20 °C
- Transportní teplota:–20 °C
- Imunogen:Full length human recombinant protein of human DTYMK(NP_036277) produced in HEK293T cell.
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 μg
- Bal.:100 µG
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100
Type: Primary
Antigen: DTYMK
Clonality: Monoclonal
Clone: 1D2
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human