Order Entry
Czech Republic
ContactUsLinkComponent
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 2B7]
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 2B7]
  ORIGCF505742
 :  OriGene
Anti-MAP2K1 Mouse Monoclonal Antibody [clone: 2B7]
  ORIGCF505742
 :  OriGene

 

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    MAP2K1
  • Klonovalita:
    Monoclonal
  • Klon:
    2B7
  • Spojení:
    Unconjugated
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • ImunoFluorescence:
    Yes
  • Izotyp:
    IgG1
  • Reaktivita:
    Monkey,
    Human,
    Dog,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    Full length
  • Křížová adsorbce:
    No
  • Formulář:
    Lyophilized
  • Skladovací pufr:
    PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
  • Molekulární hmotnost:
    43.3 kDa
  • Skladovací teplota:
    –20 °C
  • Transportní teplota:
    –20 °C
  • Imunogen:
    Full length human recombinant protein of human MAP2K1(NP_002746) produced in HEK293T cell.
  • Čištění:
    purified by affinity chromatography
  • Rozměry protilátky:
    100 μg
  • Bal.:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1:200-4000; Immunochemistry: 1:150; Immunofluorescence: 1:100

Type: Primary
Antigen: MAP2K1
Clonality: Monoclonal
Clone: 2B7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Dog, Human, Monkey, Mouse, Rat