Anti-Serpinh1 Rabbit Polyclonal Antibody
Katalogové číslo ORIGTA318915
Dodavatel: OriGene
Specifikace
- Druh protilátky:Primární
- Název antigenu:Serpin peptidase inhibitor, clade H, member 1
- Symbol antigenu:SERPINH1
- Klonovalita:Polyclonal
- Spojení:Unconjugated
- Hostitel:Rabbit
- Izotyp:IgG
- Reaktivita:Chicken,Human,Rat,Mouse
- Western blot:Yes
- Křížová adsorbce:No
- Formulář:Liquid
- Skladovací pufr:100 µg (0,2 mg/ml) Protein A purified rabbit polyclonal antibody in phosphate-buffered saline (PBS) containing 30% glycerol, 0,5% BSA, and 0,01% thimerosal.
- Skladovací teplota:–20 °C
- Transportní teplota:–20 °C
- Imunogen:Synthetic peptide surrounding amino acid 407 of human HSP47
- Čištění:purified by affinity chromatography
- Rozměry protilátky:100 μg
- Bal.:100 µG
Specifikace
O tomto produktu
Recommended Dilutions: Western Blot: 0.5-4 ?g/ml; Immunochemistry: 2.5?g/ml; Immunoprecipitation: 20?g/ml
Type: Primary
Antigen: Serpinh1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Rat