Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-Creb1 Rabbit Polyclonal Antibody
  ORIGTA318905
 :  OriGene
Anti-Creb1 Rabbit Polyclonal Antibody
  ORIGTA318905
 :  OriGene

 

  • Druh protilátky:
    Primární
  • Symbol antigenu:
    Creb1
  • Klonovalita:
    Polyclonal
  • Spojení:
    Unconjugated
  • Hostitel:
    Rabbit
  • Izotyp:
    IgG
  • Reaktivita:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Křížová adsorbce:
    No
  • Formulář:
    Liquid
  • Skladovací pufr:
    100 µg (0,5 mg/ml) purified rabbit polyclonal antibody in phosphate-buffered saline (PBS) containing 30% glycerol, 0,5% BSA and 0,01% thimerosal.
  • Skladovací teplota:
    –20 °C
  • Transportní teplota:
    –20 °C
  • Imunogen:
    Synthetic peptide surrounding amino acid 109 of rat CREB
  • Čištění:
    purified by affinity chromatography
  • Rozměry protilátky:
    100 μg
  • Bal.:
    100 µG

 

 

Recommended Dilutions: Western Blot: 0.5-4?g/ml; Immunochemistry: 5?g/ml

Type: Primary
Antigen: Creb1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat