Bottle top vacuum filtration systems
Catalog # WITG8.217.302.K
Supplier: witeg Labortechnik
Specifications
- Kapacita:300 ml
- Popis:Glass filtration system with screw cap, collecting cap, hose connector, and seal
- Ø membrány:47 mm
- Bal.:1
Specifications
About this item
The borosilicate glass vacuum filtration system with glass frit is complete with perforated PP screw thread cap GL 14, collecting flask 1000 ml, glass lower part with plastic hose connector, and seal elements PTFE.